Merck
Characterization of the enhanced apoptotic response to azidothymidine by pharmacological inhibition of NF-kB.
Life sciences (2015-03-07)
Claudia Matteucci, Antonella Minutolo, Francesca Marino-Merlo, Sandro Grelli, Caterina Frezza, Antonio Mastino, Beatrice Macchi
ABSTRAKT
The present study addresses the issue of enhanced apoptotic response to AZT following co-treatment with an NF-kB inhibitor. To investigate this issue, different cell lines were assayed for susceptibility to AZT-mediated apoptosis without or with the addition of the NF-kB inhibitor Bay-11-7085. For further investigation, U937 cells were selected as good-responder cells to the combination treatment with 32 or 128 μM AZT, and 1 μM Bay-11-7085. Inhibition of NF-kB activation by Bay-11-7085 in cells treated with AZT was assayed through Western blot analysis of p65 expression and by EMSA. Involvement of the mitochondrial pathway of apoptosis in mechanisms underlying the improved effect of AZT following Bay-11-7085 co-treatment, was evaluated by assaying the cytochrome c release and the mitochondrial membrane potential (MMP) status using the JC-1 dye. Moreover, the transcriptional activity of both anti- and pro-apoptotic genes in U937 cells after combination treatment was quantitatively evaluated through real-time PCR. We found that the combined treatment induced high levels of cytochrome c release and of MMP collapse in association with evident changes in the expression of both anti- and pro-apoptotic genes of the Bcl-2 family. Overexpression of Bcl-2 significantly suppressed the sensitization of U937 cells to an enhanced apoptotic response to AZT following co-treatment with the NF-kB inhibitor. The new findings suggest that a combination regimen based on AZT plus an NF-kB inhibitor could represent a new chemotherapeutic tool for retrovirus-related pathologies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Triton X-100, laboratory grade