Merck
Altered expression of the PLAGL1 (ZAC1/LOT1) gene in colorectal cancer: Correlations to the clinicopathological parameters.
International journal of oncology (2015-07-03)
Anna E Kowalczyk, Bartlomiej E Krazinski, Janusz Godlewski, Jolanta Kiewisz, Przemyslaw Kwiatkowski, Agnieszka Sliwinska-Jewsiewicka, Jacek Kiezun, Piotr M Wierzbicki, Gabriel Bodek, Marian Sulik, Zbigniew Kmiec
ABSTRAKT
Pleomorphic adenoma gene-like 1 gene (PLAGL1) encodes a zinc-finger nuclear transcription factor which promotes apoptosis and cell cycle arrest. Loss or downregulation of its expression has been observed in various human neoplasms. This study compared PLAGL1 expression in colorectal cancer (CRC) tissue and colon mucosa of healthy subjects at the mRNA and protein levels, and estimated its prognostic value. The PLAGL1 mRNA levels were also determined in CRC cell lines. We collected paired tumor tissue and unchanged mucosa of the large intestine from 121 CRC patients as well as 72 colon biopsies of healthy subjects obtained during screening colonoscopy. PLAGL1 mRNA levels were determined by quantitative PCR, while PLAGL1 protein expression was estimated by western blotting and immunohistochemistry. PLAGL1 mRNA level in tumor tissue was ~2-fold lower than in samples of corresponding unchanged tissues and biopsies of healthy colon mucosa. Downregulated expression of PLAGL1 mRNA was also observed in all tested CRC cell lines. Although the average content of PLAGL1 protein did not differ significantly between tumor and unchanged tissues of CRC patients or colon mucosa of healthy individuals, the decreased PLAGL1 protein levels in tumor specimens correlated with lymph node involvement, the presence of metastases and higher TNM disease stage. The PLAGL1 expression level did not correlate significantly with patient overall survival; however, the hazard ratio for patients whose tumor tissues showed reduced PLAGL1 immunohistochemical staining was twice higher than in patients with increased PLAGL1 immunoreactivity. In conclusion, these results suggest that dysregulation of PLAGL1 expression may be involved to some extent in the progression of CRC, but the so far collected patient survival data do not confirm applicability of the PLAGL1 expression level as a prognostic factor in CRC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder