Merck
Bioaccessibility of Copper in Turkish Hazelnuts (Corylus avellana L.) by Chemical Fractionation and In Vitro Methods.
Biological trace element research (2015-03-06)
Umran Seven Erdemir, Seref Gucer
ABSTRAKT
Copper is known as either an essential or toxic trace element in foods to living organisms at sufficient or excess intake levels, respectively. In this study, in vitro simulated gastric and intestinal digestion methods combined with n-octanol extraction and activated carbon adsorption were applied to Turkish hazelnuts (Corylus avellana L.) to evaluate the bioaccessibility of copper. Additionally, analytical fractionation procedures based on water, diethyl ether, n-hexane, and methanol extractions were applied to identify copper-related matrix structures. The macrochemical compositions of the samples were determined to be associated with the fractionation results. The total contents of copper in the samples, digests, and fractions were determined by inductively coupled plasma mass spectrometry after microwave-assisted digestion, and additional validation was performed using atomic absorption spectroscopy. The limits of detection and quantification were 0.016 and 0.053 mg kg(-1), respectively. The bioaccessibility of copper was found to be in the range from 16 to 31 % for hazelnut samples. The recommended dietary allowance of copper for adults is 0.9 mg daily, and considering this value, 100 g of hazelnuts supplies 33-44 % of daily copper intake.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Bile extract porcine
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%