Merck
Synthesis and characterization of a new Peptide prodrug of glucosamine with enhanced gut permeability.
PloS one (2015-05-16)
Hamed Gilzad Kohan, Kamaljit Kaur, Fakhreddin Jamali
ABSTRAKT
The aim of this study was to synthesize a peptide prodrug of glucosamine (GlcN) with increased gut permeability through the gut peptide transporter 1 (PepT1). Glycine-Valine ester derivative of GlcN (GVG) was synthesised using solid phase synthesis followed by characterization and evaluation of its physicochemical and intestinal stability. In addition, GVG was evaluated for its ability to be biotransformed to GlcN in the liver homogenate. In vitro absorption of the new prodrug through everted rat gut was also assessed. GVG demonstrated significant and meaningful increased gut permeability as compared with GlcN. It showed favorable stability in the gut and a quick cleavage to GlcN after exposure to the liver homogenate. In conclusion, a novel prodrug of glucosamine with superior gut permeability compared to GlcN was developed and successfully tested in vitro.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Fmoc chloride, BioReagent, ≥99.0% (HPLC)
Sigma-Aldrich
Fmoc chloride, 97%
Sigma-Aldrich
4-Methylmorpholine, ReagentPlus®, 99%
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
4-Methylmorpholine, purified by redistillation, ≥99.5%
Sigma-Aldrich
4-Methylmorpholine, BioXtra, suitable for protein sequencing, ≥99.5% (GC)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate, 97%
Sigma-Aldrich
Gly-Sar
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
1-Adamantylamine, 97%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC