Merck
  • Accumulation of cell-penetrating peptides in large unilamellar vesicles: A straightforward screening assay for investigating the internalization mechanism.
Accumulation of cell-penetrating peptides in large unilamellar vesicles: A straightforward screening assay for investigating the internalization mechanism.
Biopolymers (2015-04-08)
Jean-Marie Swiecicki, Margherita Di Pisa, Fabienne Burlina, Pascaline Lécorché, Christelle Mansuy, Gérard Chassaing, Solange Lavielle
ABSTRAKT
The internalization of cell-penetrating peptides (CPPs) into liposomes (large unilamellar vesicles, LUVs) was studied with a rapid and robust procedure based on the quenching of a small fluorescent probe, 7-nitrobenz-2-oxa-1,3-diazole (NBD). Quenching can be achieved by reduction with dithionite or by pH jump. LUVs with different compositions of phospholipids (PLs) were used to screen the efficacy of different CPPs. In order to "validate" the composition of the membrane models, a control cationic peptide, which does not enter eukaryotic cells, was included in the study. It was found that pure DOPG or DOPG within ternary mixtures with cholesterol are the most appropriate models for studying CPP translocation. An anionic lipid, such as DOPG, is required for the adsorption of the basic peptides on the surface of LUVs. In addition, it acts as transfer agent through the lipid bilayer. A fluid phase and/or the presence of phase defects also appear mandatory for the internalization to occur. The neutralization of charges within an inverted micelle demonstrated in the case of DOPG and also proposed for a ternary mixture of PLs might not be the only mechanism for the CPP translocation. Finally, it is shown that oleic acid facilitates the entry inside LUVs in gel phase of a series of cationic peptides including CPPs and also the negative control peptide PKCi.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Boc-Gly-OH, ≥99.0% (T)
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
DCC, 1.0 M in methylene chloride