Merck
Tonic zinc inhibits spontaneous firing in dorsal cochlear nucleus principal neurons by enhancing glycinergic neurotransmission.
Neurobiology of disease (2015-03-23)
Tamara Perez-Rosello, Charles T Anderson, Cindy Ling, Stephen J Lippard, Thanos Tzounopoulos
ABSTRAKT
In many synapses of the CNS, mobile zinc is packaged into glutamatergic vesicles and co-released with glutamate during neurotransmission. Following synaptic release, the mobilized zinc modulates ligand- and voltage-gated channels and receptors, functioning as an inhibitory neuromodulator. However, the origin and role of tonic, as opposed to phasically released, zinc are less well understood. We investigated tonic zinc in the dorsal cochlear nucleus (DCN), a zinc-rich, auditory brainstem nucleus. Our results show that application of a high-affinity, extracellular zinc chelator (ZX1) enhances spontaneous firing in DCN principal neurons (fusiform cells), consistent with inhibition of this neuronal property by tonic zinc. The enhancing effect was prevented by prior application of strychnine, a glycine receptor antagonist, suggesting that ZX1 interferes with zinc-mediated modulation of spontaneous glycinergic inhibition. In particular, ZX1 decreased the amplitude and the frequency of glycinergic miniature inhibitory postsynaptic currents in fusiform cells, from which we conclude that tonic zinc enhances glycinergic inhibitory neurotransmission. The observed zinc-mediated inhibition in spontaneous firing is present in mice lacking the vesicular zinc transporter (ZnT3), indicating that non-vesicular zinc inhibits spontaneous firing. Noise-induced increase in the spontaneous firing of fusiform cells is crucial for the induction of tinnitus. In this context, tonic zinc provides a powerful break of spontaneous firing that may protect against pathological run-up of spontaneous activity in the DCN.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Informacje o cenach i dostępności nie są obecnie dostępne.