Merck
  • Phosphate alteration of chloride behavior at the boehmite-water interface: New insights from ion-probe flow adsorption microcalorimetry.
Phosphate alteration of chloride behavior at the boehmite-water interface: New insights from ion-probe flow adsorption microcalorimetry.
Journal of colloid and interface science (2015-06-10)
S Adrian Gale, Omar R Harvey, R Dean Rhue
ABSTRAKT
Surface complexation of phosphate to aluminum oxyhydroxides can alter surface reactivity depending on the time-scale and mode of attachment. The effects of phosphate adsorption on reactivity of boehmite (γ-AlOOH) particles were investigated using ion-probe flow adsorption microcalorimetry (ipFAMC). Consistent with previous studies on adsorption energetics, probing the surface of pristine γ-AlOOH with chloride ions yielded endothermically unimodal temperature signals with a measured molar heat of exchange (ΔH(exc)) of -3.1 kJ/mol. However, when the surface of γ-AlOOH was probed with chloride following phosphate complexation, significant changes in surface reactivity resulted. Irrespective of phosphate loading, the typical endothermic response of the chloride-surface hydroxyl interaction was replaced with a multi-modal energy signature consisting of exothermic and endothermic features. These features indicate that in the presence of phosphate, the overall nature of the interaction of chloride with specific surface hydroxyls located on different exposed planes and their subsequent reactivity was transformed to a more complex environment accompanied by two or more short-lived secondary reactions. It was also shown that phosphate-promoted surface alteration of γ-AlOOH was highly selective to probing with chloride since no changes in reactivity were observed when nitrate was employed as the primary ion probe under identical experimental conditions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O