Merck
  • LY395756, an mGluR2 agonist and mGluR3 antagonist, enhances NMDA receptor expression and function in the normal adult rat prefrontal cortex, but fails to improve working memory and reverse MK801-induced working memory impairment.
LY395756, an mGluR2 agonist and mGluR3 antagonist, enhances NMDA receptor expression and function in the normal adult rat prefrontal cortex, but fails to improve working memory and reverse MK801-induced working memory impairment.
Experimental neurology (2015-09-06)
Meng-Lin Li, Sha-Sha Yang, Bo Xing, Brielle R Ferguson, Yelena Gulchina, Yan-Chun Li, Feng Li, Xi-Quan Hu, Wen-Jun Gao
ABSTRAKT
Targeting group II metabotropic glutamate receptors (mGluR2/3) has been proposed to correct the dysfunctional glutamatergic system, particularly NMDA receptor (NMDAR) hypofunction, for treatment of schizophrenia. However, how activation of mGluR2/3 affects NMDAR function in adult animals remains elusive. Here we show the effects of LY395756 (LY39), a compound acting as both an mGluR2 agonist and mGluR3 antagonist, on the NMDAR expression and function of normal adult rat prefrontal cortex (PFC) as well as working memory function in the MK801 model of schizophrenia. We found that in vivo administration of LY39 significantly increased the total protein levels of NMDAR subunits and NR2B phosphorylationin the PFC, along with the amplitude of NMDAR-mediated miniature excitatory postsynaptic currents (mEPSC) in the prefrontal cortical neurons. Moreover, LY39 also significantly increased mTOR and pmTOR expression, but not ERK1/2, Akt, and GSK3β, suggesting an activation of mTOR signaling. Indeed, the mTOR inhibitor rapamycin, and actinomycin-D, a transcription inhibitor, blocked the enhanced effects of LY39 on NMDAR-mEPSCs. These results indicate that LY39 regulates NMDAR expression and function through unidentified mTOR-mediated protein synthesis in the normal adult rat PFC. However, this change is insufficient to affect working memory function in normal animals, nor to reverse the MK801-induced working memory deficit. Our data provide the first evidence of an in vivo effect of a novel compound that acts as both an mGluR2 agonist and mGluR3 antagonist on synaptic NMDAR expression and function in the adult rat PFC, although its effect -on PFC-dependent cognitive function remains to be explored.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Przeciwciało anty-fosfo-NR2B (Ser1303), Upstate®, from rabbit
Sigma-Aldrich
Przeciwciało anty-NR2B, klon BWJHL, clone BWJHL, Upstate®, from mouse
Sigma-Aldrich
Przeciwciało anty-NR2A, klon A12W, królicze monoklonalne, culture supernatant, clone A12W, Upstate®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)