Merck
Data-independent-acquisition mass spectrometry for identification of targeted-peptide site-specific modifications.
Analytical and bioanalytical chemistry (2015-06-25)
Caleb J Porter, Michael S Bereman
ABSTRAKT
We present a novel strategy based on data-independent acquisition coupled to targeted data extraction for the detection and identification of site-specific modifications of targeted peptides in a completely unbiased manner. This method requires prior knowledge of the site of the modification along the peptide backbone from the protein of interest, but not the mass of the modification. The procedure, named multiplex adduct peptide profiling (MAPP), consists of three steps: 1) A fragment-ion tag is extracted from the data, consisting of the b-type and y-type ion series from the N and C-terminus, respectively, up to the amino-acid position that is believed to be modified; 2) MS1 features are matched to the fragment-ion tag in retention-time space, using the isolation window as a pre-filter to enable calculation of the mass of the modification; and 3) modified fragment ions are overlaid with the unmodified fragment ions to verify the mass calculated in step 2. We discuss the development, applications, and limitations of this new method for detection of unknown peptide modifications. We present an application of the method in profiling adducted peptides derived from abundant proteins in biological fluids with the ultimate objective of detecting biomarkers of exposure to reactive species.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Cerium(IV) oxide-samarium doped, <0.5 μm particle size, powder, contains 20 mol % samarium as dopant
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
SAFC
Sodium deoxycholate
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
SAFC
Iodoacetamide
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis