Merck
Isolation, characterization and investigation of Cordia dichotoma fruit polysaccharide as a herbal excipient.
International journal of biological macromolecules (2014-12-03)
Harshal Ashok Pawar, Pravin Jadhav
ABSTRAKT
The objective of the present research work was to isolate, purify and characterize Cordia dichotoma gum and investigate its disintegration property in oral tablets. The isolated gum was tested for physicochemical characteristics such as solubility, pH (1% w/w in water), swelling index, loss on drying, ash value, bulk and tapped density, Carr's index, Hausner's ratio and angle of repose. The Orodispersible tablets of valsartan were prepared by direct compression method and evaluated for average weight (mg), drug content (%), thickness (mm), hardness (kg/cm(2)), friability (%), wetting time (sec), water absorption ratio (%) and disintegration time (sec). FTIR studies revealed that there was no interaction between drug, gum and other excipients used in the study. The F4 batch with disintegration time 26.34 ± 0.78 s and in vitro release 99.64 ± 0.43% was selected as optimized formulation. This formulation was compared with conventional marketed formulation and was found superior. Batch F4 was subjected to stability studies for three months and was tested for its disintegration time, drug contents and dissolution behaviour. Batch F4 was found stable for three months at accelerated temperature.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
Magnesium stearate, technical grade
Sigma-Aldrich
Valsartan, ≥98% (HPLC)
Sigma-Aldrich
Asp-Phe methyl ester, ≥98%