Merck
Cyclic AMP Mimics the Anti-ageing Effects of Calorie Restriction by Up-Regulating Sirtuin.
Scientific reports (2015-07-15)
Zhuoran Wang, Lu Zhang, Yaru Liang, Chi Zhang, Zhiyu Xu, Lang Zhang, Ryosuke Fuji, Wei Mu, Liyuan Li, Junjun Jiang, Yong Ju, Zhao Wang
ABSTRAKT
Cyclic adenosine monophosphate (cAMP) plays an important role in many biological processes as a second messenger, and cAMP treatment has been reported to extend the lifespan of wild-type Drosophila melanogaster. Our study showed that exogenous cAMP improved ageing-related phenotypes by increasing the protein level of Sirtuins, which prevented metabolic disorders to mimic the effect of calorie restriction. Experiments in vitro showed that cAMP directly bound to SIRT1 and SIRT3 and consequently increased their activity. These findings suggest that cAMP slows the ageing process and is a good candidate to mimic calorie restriction. Our research provides a promising therapeutic strategy to target metabolic disorder-induced ageing-related diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)