Merck
OsWS1 involved in cuticular wax biosynthesis is regulated by osa-miR1848.
Plant, cell & environment (2015-05-28)
Kuaifei Xia, Xiaojin Ou, Chunzhi Gao, Huadan Tang, Yongxia Jia, Rufang Deng, Xinlan Xu, Mingyong Zhang
ABSTRAKT
Cuticular wax forms a hydrophobic layer covering aerial plant organs and acting as a protective barrier against biotic and abiotic stresses. Compared with well-known wax biosynthetic pathway, molecular regulation of wax biosynthesis is less known. Here, we show that rice OsWS1, a member of the membrane-bound O-acyl transferase gene family, involved in wax biosynthesis and was regulated by an osa-miR1848. OsWS1-tagged green fluorescent protein localized to the endoplasmic reticulum (ER). Compared with wild-type rice, OsWS1 overexpression plants displayed a 3% increase in total wax, especially a 35% increase in very long-chain fatty acids, denser wax papillae around the stoma, more cuticular wax crystals formed on leaf and stem surfaces, pollen coats were thicker and more seedlings survived after water-deficit treatment. In contrast, OsWS1-RNAi and osa-miR1848 overexpression plants exhibited opposing changes. Gene expression analysis showed that overexpression of osa-miR1848 down-regulated OsWS1 transcripts; furthermore, expression profiles of OsWS1 and osa-miR1848 were inversely correlated in the leaf, panicle and stem, and upon water-deficit treatment. These results suggest that OsWS1 is regulated by osa-miR1848 and participates in cuticular wax formation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Nonadecanoic acid, ≥98% (GC)
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Tetracosane, 99%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O