Merck
Step-Index Optical Fiber Made of Biocompatible Hydrogels.
Advanced materials (Deerfield Beach, Fla.) (2015-06-06)
Myunghwan Choi, Matjaž Humar, Seonghoon Kim, Seok-Hyun Yun
ABSTRAKT
A biocompatible step-index optical fiber made of poly(ethylene glycol) and alginate hydrogels is demonstrated. The fabricated fiber exhibits excellent light-guiding efficiency in biological tissues. Moreover, the core of hydrogel fibers can be easily doped with functional molecules and nanoparticles for localized light emission, sensing, and therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
2-Hydroxy-2-methylpropiophenone, 97%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O