Merck
  • Microemulsion liquid chromatographic method for simultaneous separation and determination of six flavonoids of Apocynum venetum leaf extract.
Microemulsion liquid chromatographic method for simultaneous separation and determination of six flavonoids of Apocynum venetum leaf extract.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-05-30)
Rui juan Song, Jun Zhou
ABSTRAKT
A simple, cost-effective, and efficient method was developed for the rapid simultaneous separation and determination of six flavonoids (rutin, hyperoside, quercetin-3-O-sophoroside, isoquercitrin, astragalin and quercetin) of Apocynum venetum leaf extract by reversed phase high performance liquid chromatography using a microemulsion system mixture as the mobile phase. Separations were performed on the Zorbax Extend-C18 column with UV detection at 360nm. The flow rate was 0.8mLmin(-1). The optimized microemulsion mobile phase consisted of 2.5% (v/v) n-butanol, 1.2% (v/v) of Genapol X-080, 0.5% (v/v) ethyl acetate and 95.8% (w/v) of aqueous 20mM phosphoric acid, pH adjusted to 6.0 with 0.3% triethylamine. Under the optimized conditions, the calibration curve for six flavonoids was linear in the range of 5-1000μgmL(-1) with the correlation coefficients greater than 0.9994. The intra-day and inter-day precision (RSD) were below 8.11% and the limits of detection (LOD) for the six flavonoids were 1.7-6.0μgmL(-1) (S/N=3). The microemulsion liquid chromatography (MELC) method was successfully applied to separate and determine the six flavonoids of A. venetum leaf extract.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Quercetin 3-D-galactoside, ≥97.0% (HPLC)
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis