Merck
Biogenic Synthesis of Fluorescent Carbon Dots at Ambient Temperature Using Azadirachta indica (Neem) gum.
Journal of fluorescence (2015-07-01)
Chinmay Phadke, Ashmi Mewada, Roopa Dharmatti, Mukeshchand Thakur, Sunil Pandey, Madhuri Sharon
ABSTRAKT
Synthesis of fluorescent Carbon Dots (CDs) from various carbonaceous materials apparently has acquired lots of interest amongst researchers as the corollary of the properties of CDs; which are subsequently getting unveiled. In this study we report the use of Azadirachta indica (Neem) Gum as a novel natural pre-cursor for synthesis of CDs at room temperature. Water soluble CDs of around 5-8 nm were obtained after treatment of the gum with ethanol and NaOH. These CDs exhibited green fluorescence in UV-light (λ = 365 nm). These CDs were found to be stable, having many bio-linkers attached on their surface, making it suitable for drug attachment and hence can serve as potential candidates for applications like drug delivery vehicles as well as for biosensors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O