Merck
  • Ribosomal Proteins RPS11 and RPS20, Two Stress-Response Markers of Glioblastoma Stem Cells, Are Novel Predictors of Poor Prognosis in Glioblastoma Patients.
Ribosomal Proteins RPS11 and RPS20, Two Stress-Response Markers of Glioblastoma Stem Cells, Are Novel Predictors of Poor Prognosis in Glioblastoma Patients.
PloS one (2015-10-28)
William H Yong, Maryam Shabihkhani, Donatello Telesca, Shuai Yang, Jonathan L Tso, Jimmy C Menjivar, Bowen Wei, Gregory M Lucey, Sergey Mareninov, Zugen Chen, Linda M Liau, Albert Lai, Stanley F Nelson, Timothy F Cloughesy, Cho-Lea Tso
ABSTRAKT
Glioblastoma stem cells (GSC) co-exhibiting a tumor-initiating capacity and a radio-chemoresistant phenotype, are a compelling cell model for explaining tumor recurrence. We have previously characterized patient-derived, treatment-resistant GSC clones (TRGC) that survived radiochemotherapy. Compared to glucose-dependent, treatment-sensitive GSC clones (TSGC), TRGC exhibited reduced glucose dependence that favor the fatty acid oxidation pathway as their energy source. Using comparative genome-wide transcriptome analysis, a series of defense signatures associated with TRGC survival were identified and verified by siRNA-based gene knockdown experiments that led to loss of cell integrity. In this study, we investigate the prognostic value of defense signatures in glioblastoma (GBM) patients using gene expression analysis with Probeset Analyzer (131 GBM) and The Cancer Genome Atlas (TCGA) data, and protein expression with a tissue microarray (50 GBM), yielding the first TRGC-derived prognostic biomarkers for GBM patients. Ribosomal protein S11 (RPS11), RPS20, individually and together, consistently predicted poor survival of newly diagnosed primary GBM tumors when overexpressed at the RNA or protein level [RPS11: Hazard Ratio (HR) = 11.5, p<0.001; RPS20: HR = 4.5, p = 0.03; RPS11+RPS20: HR = 17.99, p = 0.001]. The prognostic significance of RPS11 and RPS20 was further supported by whole tissue section RPS11 immunostaining (27 GBM; HR = 4.05, p = 0.01) and TCGA gene expression data (578 primary GBM; RPS11: HR = 1.19, p = 0.06; RPS20: HR = 1.25, p = 0.02; RPS11+RPS20: HR = 1.43, p = 0.01). Moreover, tumors that exhibited unmethylated O-6-methylguanine-DNA methyltransferase (MGMT) or wild-type isocitrate dehydrogenase 1 (IDH1) were associated with higher RPS11 expression levels [corr (IDH1, RPS11) = 0.64, p = 0.03); [corr (MGMT, RPS11) = 0.52, p = 0.04]. These data indicate that increased expression of RPS11 and RPS20 predicts shorter patient survival. The study also suggests that TRGC are clinically relevant cells that represent resistant tumorigenic clones from patient tumors and that their properties, at least in part, are reflected in poor-prognosis GBM. The screening of TRGC signatures may represent a novel alternative strategy for identifying new prognostic biomarkers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trimethylaluminum, 97%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trimesic acid, 95%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Informacje o cenach i dostępności nie są obecnie dostępne.