Merck
Microwave-assisted facile synthesis of a new tri-block chitosan conjugate with improved mucoadhesion.
Carbohydrate polymers (2015-06-17)
Ravindra V Badhe, Rabindra K Nanda, Dharmesh R Chejara, Yahya E Choonara, Pradeep Kumar, Lisa C du Toit, Viness Pillay
ABSTRAKT
A new chitosan-based tri-block conjugate, O-PEG-chitosan-N-cysteine was synthesized using microwave irradiation. For synthesis of this derivative, chitosan was modified to a PEG-chitosan conjugate followed by PEG-chitosan-cysteine using 6-O PEGylation and 2-N-thiolation, respectively. The synthesized derivative was characterized using various analytical techniques such as FT-IR and (1)H NMR spectroscopy. The conjugate was also analyzed for its biochemical, biodegradation and mucoadhesive properties. The modified chitosan conjugate exhibited improved mucoadhesion behavior (14.0 h) with greater biodegradation compared to the parent polymer (6.3h). The in silico modeling corroborated with the in vitro study demonstrating a stable complex between mucin and O-PEG-chitosan-N-cysteine conjugate (ΔE=-60.100 kcal/mol) compared to mucin and chitosan conjugate. The synthesis proposed herein, involves the use of microwave irradiation which causes a substantial reduction in the reaction time (approximately 2.30 h) compared to conventional method (35 h).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Iodomethane-12C, ≥99.9 atom % 12C, ≥99% (CP), contains copper as stabilizer
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99.5%
Sigma-Aldrich
Phthalic anhydride, ReagentPlus®, 99%
Sigma-Aldrich
Phthalic anhydride, ACS reagent, ≥99%
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
Triphenyl phosphate, ≥99%
Sigma-Aldrich
Triphenyl phosphate solution, suitable for NMR (reference standard), 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Triphenyl phosphate solution, suitable for NMR (reference standard), 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Triphenyl phosphate solution, suitable for NMR (reference standard), 0.0485 M in acetone-d6 (99.9 atom % D)
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Iodomethane solution, 2.0 M in tert-butyl methyl ether, contains copper as stabilizer
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
SAFC
L-Cysteine
Sigma-Aldrich
Dimethyl ether, ≥99.9%