Merck
  • Preparation and evaluation of surface-bonded tricationic ionic liquid silica as stationary phases for high-performance liquid chromatography.
Preparation and evaluation of surface-bonded tricationic ionic liquid silica as stationary phases for high-performance liquid chromatography.
Journal of chromatography. A (2015-04-22)
Lizhen Qiao, Xianzhe Shi, Xin Lu, Guowang Xu
ABSTRAKT
Two tricationic ionic liquids were prepared and then bonded onto the surface of supporting silica materials through "thiol-ene" click chemistry as new stationary phases for high-performance liquid chromatography. The obtained columns of tricationic ionic liquids were evaluated respectively in the reversed-phase liquid chromatography (RPLC) mode and hydrophilic interaction liquid chromatography (HILIC) mode, and possess ideal column efficiency of 80,000 plates/m in the RPLC mode with naphthalene as the test solute. The tricationic ionic liquid stationary phases exhibit good hydrophobic and shape selectivity to hydrophobic compounds, and RPLC retention behavior with multiple interactions. In the HILIC mode, the retention and selectivity were evaluated through the efficient separation of nucleosides and bases as well as flavonoids, and the typical HILIC retention behavior was demonstrated by investigating retention changes of hydrophilic solutes with water volume fraction in mobile phase. The results show that the tricationic ionic liquid columns possess great prospect for applications in analysis of hydrophobic and hydrophilic samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3,5-Tris(bromomethyl)benzene, 97%
Sigma-Aldrich
Daidzin, ≥95.0% (HPLC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Bis(trifluoromethane)sulfonimide lithium salt, 99.95% trace metals basis
Sigma-Aldrich
Bis(trifluoromethane)sulfonimide lithium salt
Sigma-Aldrich
Bis(trifluoromethane)sulfonimide lithium salt, puriss., ≥99.0% (19F-NMR)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Prunetin, ≥98.0% (TLC)
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
Pelargonidin chloride
Sigma-Aldrich
Formononetin, ≥99.0% (TLC)
Sigma-Aldrich
Naringenin, natural (US), 98%
Sigma-Aldrich
1-Vinylimidazole, ≥99%
Sigma-Aldrich
Hesperetin, ≥95%
Sigma-Aldrich
(3-Mercaptopropyl)trimethoxysilane, 95%
Sigma-Aldrich
Chrysin, ≥96.5%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Sigma-Aldrich
Phloretin, ≥99%
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Naringin, ≥90% (HPLC), from citrus fruit
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis