Merck
Competitive Homogeneous Immunoassay for Rapid Serodiagnosis of Hantavirus Disease.
Journal of clinical microbiology (2015-05-15)
Satu Hepojoki, Juuso Rusanen, Jussi Hepojoki, Visa Nurmi, Antti Vaheri, Åke Lundkvist, Klaus Hedman, Olli Vapalahti
ABSTRAKT
In this study, we describe a competitive homogeneous immunoassay that makes use of Förster resonance energy transfer (FRET) in rapid detection of pathogen-specific antibodies. The assay principle is based on competition between a monoclonal antibody (MAb) and serum antibodies to a given antigen. In the assay, named competitive FRET immunoassay (CFRET-IA), the FRET signal is induced if MAb carrying a donor label binds to an acceptor-labeled antigen. Specific antibodies in serum compete for antigen binding, resulting in reduced FRET signal. The proof-of-principle for the assay was obtained using donor-labeled Puumala virus nucleocapsid protein (PUUV-N) and acceptor-labeled anti-PUUV-N MAb. The assay was evaluated by analyzing 329 clinical samples comprising 101 from individuals with acute PUUV infection, 42 from individuals with past infection, and 186 from individuals with PUUV-seronegative sera, and the results were compared to those of reference tests. The rapid serodiagnostic test we introduced herein performed with 100% sensitivity and 99% specificity for diagnosing acute hantavirus disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)