Merck
Photolysis of Low-Brominated Diphenyl Ethers and Their Reactive Oxygen Species-Related Reaction Mechanisms in an Aqueous System.
PloS one (2015-08-15)
Mei Wang, Huili Wang, Rongbo Zhang, Meiping Ma, Kun Mei, Fang Fang, Xuedong Wang
ABSTRAKT
To date, no report was concerned with participation of reactive oxygen species in waters during photolysis of low-brominated diphenyl ethers (LBDEs). Herein, we found that electron spin resonance (ESR) signals rapidly increased with increasing irradiation time in the solution of LBDEs and 4-oxo-TMP solutions. But this phenomenon did not occur in the presence of NaN3 (1O2 quencher) demonstrating generation of 1O2 in process of LBDEs photolysis. The indirect photolytic contribution rate for BDE-47 and BDE-28 was 18.8% and 17.3% via 1O2, and 4.9% and 6.6% via ·OH, respectively. Both D2O and NaN3 experiments proved that the indirect photolysis of LBDEs was primarily attributable to 1O2. The bimolecular reaction rate constants of 1O2 with BDE-47 and BDE-28 were 3.12 and 3.64 × 106 M-1 s-1, respectively. The rate constants for BDE-47 and BDE-28 (9.01 and 17.52 × 10-3 min-1), added to isopropyl alcohol, were very close to those (9.65 and 18.42 × 10-3 min-1) in water, proving the less indirect photolytic contribution of ·OH in water. This is the first comprehensive investigation examining the indirect photolysis of LBDEs in aqueous solution.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2,2,6,6-Tetramethyl-4-piperidone, 95%
Sigma-Aldrich
4-Nitroanisole, 97%
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, ≥99%
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, 99%
Sigma-Aldrich
Diphenyl ether, ≥99%, FG
Sigma-Aldrich
Furfuryl alcohol, 98%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Furfuryl alcohol, ≥97%, FG
Sigma-Aldrich
Furfuryl alcohol, natural, ≥95%, FG