Merck
Growth Kinetics, Characterization, and Plasticity of Human Menstrual Blood Stem Cells.
Iranian journal of medical sciences (2016-03-19)
Davood Mehrabani, Roshanak Bahrami Nazarabadi, Maryam Kasraeian, Amin Tamadon, Mehdi Dianatpour, Akbar Vahdati, Shahrokh Zare, Farnaz Ghobadi
ABSTRAKT
One of the readily available sources of mesenchymal stem cells (MSCs) is menstrual blood-derived stem cells (Men-SCs), which exhibit characteristics similar to other types of MSCs. This study was performed to determine the growth kinetics, plasticity, and characterization of Men-SCs in women. During spring 2014 in the southern Iranian city of Shiraz, menstrual blood (5 mL) was obtained from 10 women on their third day of menstruation in 2 age groups of 30 to 40 and 40 to 50 years old. Ficoll was used to separate the mononuclear cell fraction. After the Men-SCs were cultured, they were subcultured up to passage 4. Growth behavior and population doubling time were evaluated by seeding 5×10(4) cells into 12- and 24-well culture plates, and the colonies were enumerated. The expression of CD44, CD90, and CD34 was evaluated. The osteogenic potential was assessed by alizarin red staining. The Men-SCs were shown to be plastic adherent and spindle-shaped. Regarding the growth curves in the 12- and 24-well culture plates, it was demonstrated that in the women aged between 30 and 40 years, population doubling time was 55.5 and 62 hours, respectively, while these values in the women aged between 40 and 50 years were 70.4 and 72.4 hours, correspondingly. Positive expression of CD44 and CD90 and negative expression of CD34 were noted. In the osteogenic differentiation medium, the cells differentiated toward osteoblasts. As human Men-SCs are easily collectable without any invasive procedure and are a safe and rapid source of MSCs, they can be a good candidate for stem cell banking and cell transplantation in women.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Demecolcine solution, 10 μg/mL in HBSS, ACF Qualified, BioXtra
Informacje o cenach i dostępności nie są obecnie dostępne.