Merck
Activity-regulated trafficking of the palmitoyl-acyl transferase DHHC5.
Nature communications (2015-09-04)
G Stefano Brigidi, Brendan Santyr, Jordan Shimell, Blair Jovellar, Shernaz X Bamji
ABSTRAKT
Synaptic plasticity is mediated by the dynamic localization of proteins to and from synapses. This is controlled, in part, through activity-induced palmitoylation of synaptic proteins. Here we report that the ability of the palmitoyl-acyl transferase, DHHC5, to palmitoylate substrates in an activity-dependent manner is dependent on changes in its subcellular localization. Under basal conditions, DHHC5 is bound to PSD-95 and Fyn kinase, and is stabilized at the synaptic membrane through Fyn-mediated phosphorylation of a tyrosine residue within the endocytic motif of DHHC5. In contrast, DHHC5's substrate, δ-catenin, is highly localized to dendritic shafts, resulting in the segregation of the enzyme/substrate pair. Neuronal activity disrupts DHHC5/PSD-95/Fyn kinase complexes, enhancing DHHC5 endocytosis, its translocation to dendritic shafts and its association with δ-catenin. Following DHHC5-mediated palmitoylation of δ-catenin, DHHC5 and δ-catenin are trafficked together back into spines where δ-catenin increases cadherin stabilization and recruitment of AMPA receptors to the synaptic membrane.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało przeciw fosfotyrozynie, klon ®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-ZDHHC5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®
Roche
Anty-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Anti-PSD-95 (Ab-2) Mouse mAb (7E3-1B8), liquid, clone 7E3-1B8, Calbiochem®
Sigma-Aldrich
Przeciwciało anty-GluR1, rekombinowane, królicze monoklonalne, Upstate®, from rabbit