Merck
  • Synthesis and structure-activity relationships of potent new angiotensin converting enzyme inhibitors containing saturated bicyclic amino acids.
Synthesis and structure-activity relationships of potent new angiotensin converting enzyme inhibitors containing saturated bicyclic amino acids.
Journal of medicinal chemistry (1987-06-01)
C J Blankley, J S Kaltenbronn, D E DeJohn, A Werner, L R Bennett, G Bobowski, U Krolls, D R Johnson, W M Pearlman, M L Hoefle
ABSTRAKT
The synthesis of a series of novel, potent angiotensin converting enzyme (ACE) inhibitors containing saturated bicyclic amino acids in place of proline is described. Octahydroindole-2-carboxylic acid, octahydroisoindole-1-carboxylic acid, and octahydro-3-oxoisoindole-1-carboxylic acid can replace proline in both sulfhydryl and non-sulfhydryl ACE inhibitors to give compounds equipotent to captopril and enalapril both in vitro and in vivo. Structure-activity relationships are discussed. Compound 11a (CI-907, indolapril) has advanced to clinical evaluation.