Merck
Interleukin-13 interferes with activation-induced t-cell apoptosis by repressing p53 expression.
Cellular & molecular immunology (2015-07-21)
Li Yang, Ling-Zhi Xu, Zhi-Qiang Liu, Gui Yang, Xiao-Rui Geng, Li-Hua Mo, Zhi-Gang Liu, Peng-Yuan Zheng, Ping-Chang Yang
ABSTRAKT
The etiology and the underlying mechanism of CD4(+) T-cell polarization are unclear. This study sought to investigate the mechanism by which interleukin (IL)-13 prevents the activation-induced apoptosis of CD4(+) T cells. Here we report that CD4(+) T cells expressed IL-13 receptor α2 in the intestine of sensitized mice. IL-13 suppressed both the activation-induced apoptosis of CD4(+) T cells and the expression of p53 and FasL. Exposure to recombinant IL-13 inhibited activation-induced cell death (AICD) along with the expression of p53, caspase 3, and tumor necrosis factor-α in CD4(+) T cells. Administration of an anti-IL-13 antibody enhanced the effect of specific immunotherapy on allergic inflammation in the mouse intestine, enforced the expression of p53 in intestinal CD4(+) T cells, and enhanced the frequency of CD4(+) T-cell apoptosis upon challenge with specific antigens. In summary, blocking IL-13 enhances the therapeutic effect of antigen-specific immunotherapy by regulating apoptosis and thereby enforcing AICD in CD4(+) T cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT6
Informacje o cenach i dostępności nie są obecnie dostępne.