Merck
The RAD51 paralogs ensure cellular protection against mitotic defects and aneuploidy.
Journal of cell science (2012-10-31)
Amélie Rodrigue, Yan Coulombe, Karine Jacquet, Jean-Phillipe Gagné, Céline Roques, Stéphane Gobeil, Guy Poirier, Jean-Yves Masson
ABSTRAKT
The interplay between homologous DNA recombination and mitotic progression is poorly understood. The five RAD51 paralogs (RAD51B, RAD51C, RAD51D, XRCC2 and XRCC3) are key enzymes for DNA double-strand break repair. In our search for specific functions of the various RAD51 paralogs, we found that inhibition of XRCC3 elicits checkpoint defects, while inhibition of RAD51B or RAD51C induces G2/M cell cycle arrest in HeLa cells. Using live-cell microscopy we show that in XRCC3-knockdown cells the spindle assembly checkpoint persists and there is a higher frequency of chromosome misalignments, anaphase bridges, and aneuploidy. We observed centrosome defects in the absence of XRCC3. While RAD51B and RAD51C act early in homologous recombination, XRCC3 functions jointly with GEN1 later in the pathway at the stage of Holliday junction resolution. Our data demonstrate that Holliday junction resolution has critical functions for preventing aberrant mitosis and aneuploidy in mitotic cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-fosfo-histonowe H3 (Ser10), marker mitozy, Upstate®, from rabbit
Sigma-Aldrich
Przeciwciało anty-PICH, klon 142-26-3, clone 142-26-3, from mouse
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid