Merck
Mechanical stimulation improves tissue-engineered human skeletal muscle.
American journal of physiology. Cell physiology (2002-10-10)
Courtney A Powell, Beth L Smiley, John Mills, Herman H Vandenburgh
ABSTRAKT
Human bioartificial muscles (HBAMs) are tissue engineered by suspending muscle cells in collagen/MATRIGEL, casting in a silicone mold containing end attachment sites, and allowing the cells to differentiate for 8 to 16 days. The resulting HBAMs are representative of skeletal muscle in that they contain parallel arrays of postmitotic myofibers; however, they differ in many other morphological characteristics. To engineer improved HBAMs, i.e., more in vivo-like, we developed Mechanical Cell Stimulator (MCS) hardware to apply in vivo-like forces directly to the engineered tissue. A sensitive force transducer attached to the HBAM measured real-time, internally generated, as well as externally applied, forces. The muscle cells generated increasing internal forces during formation which were inhibitable with a cytoskeleton depolymerizer. Repetitive stretch/relaxation for 8 days increased the HBAM elasticity two- to threefold, mean myofiber diameter 12%, and myofiber area percent 40%. This system allows engineering of improved skeletal muscle analogs as well as a nondestructive method to determine passive force and viscoelastic properties of the resulting tissue.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Horse Serum, Donor herd, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse, clone MY-32, ascites fluid
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Tropomyosin (Sarcomeric) antibody, Mouse monoclonal, clone CH1, purified from hybridoma cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.