Merck
Scratch2 modulates neurogenesis and cell migration through antagonism of bHLH proteins in the developing neocortex.
Cerebral cortex (New York, N.Y. : 1991) (2012-11-28)
Vanessa Paul, Anton B Tonchev, Kristine A Henningfeld, Evangelos Pavlakis, Barbara Rust, Tomas Pieler, Anastassia Stoykova
ABSTRAKT
Scratch genes (Scrt) are neural-specific zinc-finger transcription factors (TFs) with an unknown function in the developing brain. Here, we show that, in addition to the reported expression of mammalian Scrt2 in postmitotic differentiating and mature neurons in the developing and early postnatal brain, Scrt2 is also localized in subsets of mitotic and neurogenic radial glial (RGP) and intermediate (IP) progenitors, as well as in their descendants-postmitotic IPs and differentiating neurons at the border subventricular/intermediate zone. Conditional activation of transgenic Scrt2 in cortical progenitors in mice promotes neuronal differentiation by favoring the direct mode of neurogenesis of RGPs at the onset of neurogenesis, at the expense of IP generation. Neuronal amplification via indirect IP neurogenesis is thereby extenuated, leading to a mild postnatal reduction of cortical thickness. Forced in vivo overexpression of Scrt2 suppressed the generation of IPs from RGPs and caused a delay in the radial migration of upper layer neurons toward the cortical plate. Mechanistically, our results indicate that Scrt2 negatively regulates the transcriptional activation of the basic helix loop helix TFs Ngn2/NeuroD1 on E-box containing common target genes, including Rnd2, a well-known major effector for migrational defects in developing cortex. Altogether, these findings reveal a modulatory role of Scrt2 protein in cortical neurogenesis and neuronal migration.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-SCRT2 antibody produced in rabbit, affinity isolated antibody
Informacje o cenach i dostępności nie są obecnie dostępne.