Merck
Increases in Serum Growth Hormone Concentrations Associated with GHB Administration.
Journal of analytical toxicology (2016-10-17)
Alan D Brailsford, Christiaan Bartlett, Andrew T Kicman, David A Cowan
ABSTRAKT
The administration of gamma-hydroxybutyrate (GHB) has been reported to augment the increase in growth hormone (GH) secretion associated with the onset of sleep. The ability of GHB to stimulate GH production in the absence of sleep in both male and female volunteers was investigated as part of a GHB administration study. Twelve healthy volunteers (six men and six women) were given a small oral dose (25 mg/kg) of GHB (as Xyrem