Merck
  • Functional Teas from the Leaves of Arbutus unedo: Phenolic Content, Antioxidant Activity, and Detection of Efficient Radical Scavengers.
Functional Teas from the Leaves of Arbutus unedo: Phenolic Content, Antioxidant Activity, and Detection of Efficient Radical Scavengers.
Plant foods for human nutrition (Dordrecht, Netherlands) (2017-04-20)
Ioannis Erkekoglou, Nikolaos Nenadis, Efrosini Samara, Fani Th Mantzouridou
ABSTRAKT
The phenolic content/composition and antioxidant activity of hot/cold infusion and decoction from the leaves of Arbutus unedo were studied for the first time. 1,1-diphenyl-2-picrylhydrazyl (DPPH
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.