Merck
Efficient Production of Papillomavirus Gene Delivery Vectors in Defined In Vitro Reactions.
Molecular therapy. Methods & clinical development (2017-05-13)
Carla Cerqueira, Cynthia D Thompson, Patricia M Day, Yuk-Ying S Pang, Douglas R Lowy, John T Schiller
ABSTRAKT
Papillomavirus capsids can package a wide variety of nonviral DNA plasmids and deliver the packaged genetic material to cells, making them attractive candidates for targeted gene delivery vehicles. However, the papillomavirus vectors generated by current methods are unlikely to be suitable for clinical applications. We have developed a chemically defined, cell-free, papillomavirus-based vector production system that allows the incorporation of purified plasmid DNA (pseudogenome) into high-titer papillomavirus L1/L2 capsids. We investigated the incorporation of several DNA forms into a variety of different papillomavirus types, including human and animal types. Our results show that papillomavirus capsids can package and transduce linear or circular DNA under defined conditions. Packaging and transduction efficiencies were surprisingly variable across capsid types, DNA forms, and assembly reaction conditions. The pseudoviruses produced by these methods are sensitive to the same entry inhibitors as cell-derived pseudovirions, including neutralizing antibodies and heparin. The papillomavirus vector production systems developed in this study generated as high as 10
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6-Carboxyfluorescein, ≥96% (HPLC)
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, ≥180 USP units/mg
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Brij® 58, average Mn ~1124
Sigma-Aldrich
γ-Secretase Inhibitor XXI, Compound E, This g-secretase inhibitor, CAS 209986-17-4, is a cell-permeable, potent, selective, non-transition-state analog inhibitor of γ-secretase and Notch processing. Lowers Aβ levels in APP transgenic mice
Sigma-Aldrich
Furin Inhibitor I, Furin Inhibitor I, is a peptidyl chloromethylketone that binds to the catalytic site of furin and blocks its activity.