Merck
  • Nickel toxicity to cardinal tetra (Paracheirodon axelrodi) differs seasonally and among the black, white and clear river waters of the Amazon basin.
Nickel toxicity to cardinal tetra (Paracheirodon axelrodi) differs seasonally and among the black, white and clear river waters of the Amazon basin.
Water research (2017-06-25)
Aleicia Holland, Chris M Wood, D Scott Smith, Tiago Gabriel Correia, Adalberto L Val
ABSTRAKT
This study investigated the acute toxicity of nickel (Ni) to cardinal tetra (Paracheirodon axelrodi), within the three main water types of the Amazon basin: black (Rio Negro), white (Rio Solimões) and clear (Rio Tapajós) during the wet and dry season at pH 7 (representative of white and clear rivers) and pH 4 (representative of black waters). The influence of dissolved organic carbon (DOC) quality on Ni toxicity within the three waters was also explored via the use of DOC isolates. Differences in water chemistry, DOC quality and ion concentrations were shown between waters and between seasons. Toxicity of Ni was shown to vary between river waters, seasons, and pHs. Ni was significantly less toxic during the dry season at pH 4 in all three river waters; for example, black water during the wet season had an LC
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.