- Blocked transcription through KvDMR1 results in absence of methylation and gene silencing resembling Beckwith-Wiedemann syndrome.
Blocked transcription through KvDMR1 results in absence of methylation and gene silencing resembling Beckwith-Wiedemann syndrome.
Development (Cambridge, England) (2017-04-22)
Vir B Singh, Sirinapa Sribenja, Kayla E Wilson, Kristopher M Attwood, Joanna C Hillman, Shilpa Pathak, Michael J Higgins
PMID28428215
ABSTRAKT
The maternally methylated KvDMR1 ICR regulates imprinted expression of a cluster of maternally expressed genes on human chromosome 11p15.5. Disruption of imprinting leads to Beckwith-Wiedemann syndrome (BWS), an overgrowth and cancer predisposition condition. In the majority of individuals with BWS, maternal-specific methylation at KvDMR1 is absent and genes under its control are repressed. We analyzed a mouse model carrying a poly(A) truncation cassette inserted to prevent RNA transcripts from elongation through KvDMR1. Maternal inheritance of this mutation resulted in absence of DNA methylation at KvDMR1, which led to biallelic expression of
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Informacje o cenach i dostępności nie są obecnie dostępne.
Informacje o cenach i dostępności nie są obecnie dostępne.