Merck
Direct Activation of BAX by BTSA1 Overcomes Apoptosis Resistance in Acute Myeloid Leukemia.
Cancer cell (2017-10-11)
Denis E Reyna, Thomas P Garner, Andrea Lopez, Felix Kopp, Gaurav S Choudhary, Ashwin Sridharan, Swathi-Rao Narayanagari, Kelly Mitchell, Baoxia Dong, Boris A Bartholdy, Loren D Walensky, Amit Verma, Ulrich Steidl, Evripidis Gavathiotis
ABSTRAKT
The BCL-2 family protein BAX is a central mediator of apoptosis. Overexpression of anti-apoptotic BCL-2 proteins contributes to tumor development and resistance to therapy by suppressing BAX and its activators. We report the discovery of BTSA1, a pharmacologically optimized BAX activator that binds with high affinity and specificity to the N-terminal activation site and induces conformational changes to BAX leading to BAX-mediated apoptosis. BTSA1-induced BAX activation effectively promotes apoptosis in leukemia cell lines and patient samples while sparing healthy cells. BAX expression levels and cytosolic conformation regulate sensitivity to BTSA1. BTSA1 potently suppressed human acute myeloid leukemia (AML) xenografts and increased host survival without toxicity. This study provides proof-of-concept for direct BAX activation as a treatment strategy in AML.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Boron-11B, 95 atom % 11B
Sigma-Aldrich
Przeciwciało anty-Bak, NT, Upstate®, from rabbit
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.
Sigma-Aldrich
BTSA1, ≥98% (HPLC)