- Alisertib promotes apoptosis and autophagy in melanoma through p38 MAPK-mediated aurora a signaling.
Alisertib promotes apoptosis and autophagy in melanoma through p38 MAPK-mediated aurora a signaling.
Oncotarget (2018-01-02)
Yuan-Yuan Shang, Ming Yao, Zhi-Wei Zhou, Jian-Cui, Li-Xia, Rong-Ying Hu, Ying-Yao Yu, Qiong-Gao, Biao-Yang, Yu-Xi Liu, Jie Dang, Shu-Feng Zhou, Nan-Yu
PMID29291012
ABSTRAKT
We investigated the efficacy of Alisertib (ALS), a selective Aurora kinase A (AURKA) inhibitor, in melanoma. We found that ALS exerts anti-proliferative, pro-apoptotic, and pro-autophagic effects on A375 and skmel-5 melanoma cells by inhibiting p38 MAPK signaling. SB202190, a p38 MAPK-selective inhibitor, enhanced ALS-induced apoptosis and autophagy in both cell lines. ALS induced cell cycle arrest in melanoma cells through activation of the p53/p21/cyclin B1 pathway. Knockdown of p38 MAPK enhanced ALS-induced apoptosis and reduced ALS-induced autophagy. Inhibition of autophagy sensitized melanoma cells to ALS-induced apoptosis. These data indicate ALS is a potential therapeutic agent for melanoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Informacje o cenach i dostępności nie są obecnie dostępne.