- Peptide Epimerization Machineries Found in Microorganisms.
Peptide Epimerization Machineries Found in Microorganisms.
Frontiers in microbiology (2018-02-23)
Yasushi Ogasawara, Tohru Dairi
PMID29467749
ABSTRAKT
D-Amino acid residues have been identified in peptides from a variety of eukaryotes and prokaryotes. In microorganisms, UDP-