Skip to Content
MilliporeSigma

BDPH9BCD02CA

Bide Pharmatech

5-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one

Synonym(s):

5-chloro-4-fluoro-1-indanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H6O1Cl1F1
CAS Number:
Molecular Weight:
184.6
MDL number:


Inchi Code

1S/C9H6ClFO/c10-7-3-1-5-6(9(7)11)2-4-8(5)12/h1,3H,2,4H2

Inchi Key

VRURYMJQFZRENG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to yellow to grey powder or crystals



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library