Skip to Content
MilliporeSigma

BDPH9BCD0331

Bide Pharmatech

2-(4-Chlorobenzyl)malonic acid

Synonym(s):

2-(4-chlorobenzyl)malonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O4Cl1
CAS Number:
Molecular Weight:
228.63
MDL number:


Purity

98%

Inchi Code

1S/C10H9ClO4/c11-7-3-1-6(2-4-7)5-8(9(12)13)10(14)15/h1-4,8H,5H2,(H,12,13)(H,14,15)

Inchi Key

QNZGBKFZLKIUPN-UHFFFAOYSA-N

Shipping Temperature

Normal

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 2-(4-Chlorobenzyl)malonic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library