Inchi Code
1S/C11H18O5/c1-4-7-8(10(13)15-5-2)9(12)11(14)16-6-3/h8H,4-7H2,1-3H3
Inchi Key
FQQZGZZZSPUFKV-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under Diethyl 2-oxo-3-propylsuccinate
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
