Skip to Content
MilliporeSigma

BDPH9BCD8B4E

Bide Pharmatech

Diethyl octylphosphonate

Synonym(s):

diethyl octylphosphonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H27O3P1
CAS Number:
Molecular Weight:
250.32
MDL number:


Inchi Code

1S/C12H27O3P/c1-4-7-8-9-10-11-12-16(13,14-5-2)15-6-3/h4-12H2,1-3H3

Inchi Key

MVDVRXOWIPONFY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under Diethyl octylphosphonate


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library