Skip to Content
MilliporeSigma

BDPH9BCD8EA4

Bide Pharmatech

2-(5-Bromopyridin-2-yl)-2-methylpropanenitrile

Synonym(s):

2-(5-bromo-2-pyridinyl)-2-methylpropanenitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H9N2Br1
CAS Number:
Molecular Weight:
225.09
MDL number:


Inchi Code

1S/C9H9BrN2/c1-9(2,6-11)8-4-3-7(10)5-12-8/h3-5H,1-2H3

Inchi Key

LCFRTRIRICENNR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Colorless to Yellow Solid or Semi-solid or liquid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library