Skip to Content
MilliporeSigma

BDPH9BCD9653

Bide Pharmatech

Ethyl 3-(chloromethyl)benzoate

Synonym(s):

ethyl 3-(chloromethyl)benzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H11O2Cl1
CAS Number:
Molecular Weight:
198.65
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H11ClO2/c1-2-13-10(12)9-5-3-4-8(6-9)7-11/h3-6H,2,7H2,1H3

Inchi Key

ZJNVMXXFCNKXLT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Form: liquid / Colour: Colorless - Very pale reddish yellow - Yellow

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under Ethyl 3-(chloromethyl)benzoate



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library