Inchi Code
1S/C10H12Br2/c1-10(2,3)7-4-5-8(11)9(12)6-7/h4-6H,1-3H3
Inchi Key
LZOUMICSOKCMJT-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Colorless to yellow solid or liquid
Still not finding the right product?
Explore all of our products under 1,2-Dibromo-4-tert-butylbenzene
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
