Inchi Code
1S/C8H10N4O4/c9-11-7(15)3-1-5(13)4(2-6(3)14)8(16)12-10/h1-2,13-14H,9-10H2,(H,11,15)(H,12,16)
Inchi Key
DQZASJMFKRPEEF-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Keep in dark place, inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 2,5-Dihydroxyterephthalohydrazide
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
