Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now

BDPH9BCDBBBF

Bide Pharmatech

2,4-Dichloro-3,5-dimethylphenol

Synonym(s):

2,4-dichloro-3,5-dimethylphenol

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C8H8O1Cl2
CAS Number:
Molecular Weight:
191.06
MDL number:

Inchi Code

1S/C8H8Cl2O/c1-4-3-6(11)8(10)5(2)7(4)9/h3,11H,1-2H3

Inchi Key

IYOLBFFHPZOQGW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

92%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to light yellow-red powder or crystal

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library