Skip to Content
MilliporeSigma

BDPH9BCDCA89

Bide Pharmatech

2,3,6-Trichloro-4-(trifluoromethyl)pyridine

Synonym(s):

2,3,6-trichloro-4-(trifluoromethyl)pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H1N1Cl3F3
CAS Number:
Molecular Weight:
250.43
MDL number:


Inchi Code

1S/C6HCl3F3N/c7-3-1-2(6(10,11)12)4(8)5(9)13-3/h1H

Inchi Key

KCMZCZXPLOZOGG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library