Skip to Content
MilliporeSigma

BDPH9BCDCD36

Bide Pharmatech

6-Bromopyrido[2,3-b]pyrazine

Synonym(s):

6-bromopyrido[2,3-b]pyrazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H4N3Br1
CAS Number:
Molecular Weight:
210.03
MDL number:


Inchi Code

1S/C7H4BrN3/c8-6-2-1-5-7(11-6)10-4-3-9-5/h1-4H

Inchi Key

JLMAHBCUEYQXKS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 6-Bromopyrido[2,3-b]pyrazine




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library