Skip to Content
MilliporeSigma

BDPH9BCDCD67

Bide Pharmatech

5-Bromo-1H-indazole-3-carbaldehyde

Synonym(s):

5-bromo-1H-indazole-3-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H5O1N2Br1
CAS Number:
Molecular Weight:
225.04
MDL number:


Inchi Code

1S/C8H5BrN2O/c9-5-1-2-7-6(3-5)8(4-12)11-10-7/h1-4H,(H,10,11)

Inchi Key

ILGTYHMEQSSHFG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 5-Bromo-1H-indazole-3-carbaldehyde


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library