Inchi Code
1S/C32H24NP/c33-29-21-19-23-11-7-9-17-27(23)31(29)32-28-18-10-8-12-24(28)20-22-30(32)34(25-13-3-1-4-14-25)26-15-5-2-6-16-26/h1-22H,33H2
Inchi Key
CXPJTDITWGPSMR-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Keep in dark place, sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Explore all of our products under (1R)-2'-(Diphenylphosphino)-[1,1'-binaphthalen]-2-amine
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
