Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

BDPH9BCDD67A

Bide Pharmatech

Cyclopentanone-2-carbonitrile

Synonym(s):

2-oxocyclopentanecarbonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C6H7O1N1
CAS Number:
Molecular Weight:
109.13
MDL number:

Inchi Code

1S/C6H7NO/c7-4-5-2-1-3-6(5)8/h5H,1-3H2

Inchi Key

IPMQSLPLJDKUPI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library