Skip to Content
MilliporeSigma

BDPH9BCDDAC3

Bide Pharmatech

(S)-1,10-Bis(3,5-di-tert-butyl-4-methoxyphenyl)-12-hydroxy-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocine 12-oxide

Synonym(s):

1,10-bis(3,5-di-tert-butyl-4-methoxyphenyl)-12-hydroxy-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocine 12-oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C47H59O6P1
CAS Number:
Molecular Weight:
750.96
MDL number:


Inchi Code

1S/C47H59O6P/c1-43(2,3)33-23-29(24-34(41(33)50-13)44(4,5)6)31-17-15-27-19-21-47-22-20-28-16-18-32(40(38(28)47)53-54(48,49)52-39(31)37(27)47)30-25-35(45(7,8)9)42(51-14)36(26-30)46(10,11)12/h15-18,23-26H,19-22H2,1-14H3,(H,48,49)

Inchi Key

HLRBLHQIKBAQLS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library